Silver(I) acetate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4701-10G
93-4701-50G
100205-584EA
72.18
USD
100205-584
100202-800
Silver(I) acetate ≥99%
Silver (I) acetate
Formula:
C₂H₃AgO₂ Melting Pt: Dec. Density: 3.26 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012446 CAS Number: 563-63-3 EINECS: 209-254-9 Merck Index: 12,08648 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...