Dipotassium hexachloropalladate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassium hexachloropalladate(IV)

93-4610-5G 93-4610-1G
100207-074EA 406.22 USD
100207-074 100207-072
Dipotassium hexachloropalladate ≥99%
Dipotassium hexachloropalladate
Formula: Cl₆K₂Pd
Melting Pt: Dec.
Density: 2,738
MDL Number: MFCD00043081
CAS Number: 16919-73-6
EINECS: 240-974-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR