Molybdenum ≥99.95%, rods
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4255-50G
93-4255-250G
100204-328EA
104.74
USD
100204-328
100202-134
Molybdenum ≥99.95%, rods
Molybdenum
Formula:
Mo MW: 95.94 g/mol Boiling Pt: 3617 Melting Pt: 2622 Density: 10.2 Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...