Niobium ≥99.8%, foil

Supplier: STREM CHEMICALS INC MS
Warning

93-4135-500MM 93-4135-100MM
100196-658EA 553.42 USD
100196-658 100210-402
Niobium ≥99.8%, foil
Niobium
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR