Niobium ≥99.8%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-4132-25G 93-4132-100G
100197-854EA 99.08 USD
100197-854 100211-258
Niobium ≥99.8%, powder
Niobium
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR