Zirconium ≥99.5%, foil
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4046-100MM
93-4046-400MM
100197-852EA
519.45
USD
100197-852
100206-028
Zirconium ≥99.5%, foil
Zirconium
Formula:
Zr Boiling Pt: 3577 °C Melting Pt: 1852 °C Density: 6,506 |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 UN: 2858 ADR: 4,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...