Strontium chloride hexahydrate ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-3807-500G
93-3807-100G
100199-562EA
157.11
USD
100199-562
100205-550
Strontium chloride hexahydrate ≥99% ACS
Strontium chloride hexahydrate
Formula:
SrCl₂·6H₂O MW: 266.62 g/mol Boiling Pt: 100 °C (1013 hPa) Melting Pt: 115 °C Density: 1.93 Storage Temperature: Ambient |
MDL Number:
MFCD00149865 CAS Number: 10025-70-4 EINECS: 233-971-6 Merck Index: 13,08920 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...