Rubidium fluoride, anhydrous ≥99.8% (Rb basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-3730-25G 93-3730-5G
100197-226EA 734.58 USD
100197-226 100205-548
Rubidium fluoride, anhydrous ≥99.8% (Rb basis)
Rubidium fluoride
Formula: FRb
Boiling Pt: 1410 °C
Melting Pt: 775 °C
Density: 3.557
MDL Number: MFCD00011191
CAS Number: 13446-74-7
EINECS: 236-603-2
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR