Sodium selenite ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-3414-50G 93-3414-250G
100199-078EA 76.43 USD
100199-078 100199-076
Sodium selenite ≥99%
Sodium selenite
Formula: Na₂SeO₃
MW: 172.94 g/mol
Melting Pt: 710 °C
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003489
CAS Number: 10102-18-8
EINECS: 233-267-9
UN: 2630
ADR: 6.1,I
Merck Index: 12,08822

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR