Germanium ≥99.999%, pieces

Supplier: STREM CHEMICALS INC MS

93-3232-50G 93-3232-10G
100210-368EA 1078.52 USD
100210-368 100210-366
Germanium ≥99.999%, pieces
Germanium
Formula: Ge
Boiling Pt: 2830 °C
Melting Pt: 937.4 °C
Density: 5.323
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR