Germanium ≥99.999%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-3231-5G 93-3231-25G
100205-526EA 158.52 USD
100205-526 100209-432
Germanium ≥99.999%, powder
Germanium
Formula: Ge
Boiling Pt: 2830 °C
Melting Pt: 937.4 °C
Density: 5.323
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
UN: 3089
ADR: 4,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR