Gallium metal ≥99.9999%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-3146-50G
93-3146-10G
100202-450EA
737.42
USD
100202-450
100205-522
Gallium metal ≥99.9999%
Gallium
Formula:
Ga Boiling Pt: 2403 °C Melting Pt: 29.78 °C Density: 5.904 Storage Temperature: Ambient |
MDL Number:
MFCD00134045 CAS Number: 7440-55-3 EINECS: 231-163-8 UN: 2803 ADR: 8,III Merck Index: 12,04364 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...