Gallium metal ≥99.99%

Supplier: STREM CHEMICALS INC MS
Danger

93-3145-10G 93-3145-50G
100201-100EA 99.08 USD
100201-100 100201-102
Gallium metal ≥99.99%
Gallium
Formula: Ga
Boiling Pt: 2403 °C
Melting Pt: 29.78 °C
Density: 5.904
Storage Temperature: Ambient
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III
Merck Index: 12,04364

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR