Digallium trioxide ≥99.998%, PURATREM

Supplier: STREM CHEMICALS INC MS

93-3106-25G 93-3106-5G
100210-358EA 300.06 USD
100210-358 100196-620
Digallium trioxide ≥99.998%, PURATREM
Digallium trioxide
Formula: Ga₂O₃
MW: 187.44
Melting Pt: 1794.85
Density: 5.88
MDL Number: MFCD00011020
CAS Number: 12024-21-4
EINECS: 234-691-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR