Zinc ≥97.5%, dust

Supplier: STREM CHEMICALS INC MS
Danger

93-3056-5000G 93-3056-1000G 93-3056-250G
100202-082EA 387.82 USD
100202-082 100210-356 100205-516
Zinc ≥97.5%, dust
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3 II
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR