Zinc ≥99.99%, rods

Supplier: STREM CHEMICALS INC MS

93-3055-100G 93-3055-500G
100202-080EA 73.6 USD
100202-080 100206-026
Zinc ≥99.99%, rods
Zinc
Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR