Zinc chloride ≥99.99% (Zn basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Butter of zinc
, Zinc (II) chloride
, Zinc dichloride
, Dichlorozinc
93-3052-50G
93-3052-10G
100203-448EA
781.29
USD
100203-448
100203-446
Zinc chloride ≥99.99% (Zn basis), PURATREM
Zinc chloride
Formula:
ZnCl₂ MW: 136.3 g/mol Boiling Pt: 732 °C Melting Pt: 283 °C Density: 2.91 Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 12,10261 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...