Iridium, black ≥99.9% (by IR)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
77-2000-500MG
77-2000-2G
100205-194EA
317.05
USD
100205-194
100200-716
Iridium, black ≥99.9% (by IR)
Iridium
Formula:
Ir Boiling Pt: 4130 °C Melting Pt: 2410 °C Density: 22.421 |
MDL Number:
MFCD00011062 CAS Number: 7439-88-5 EINECS: 231-095-9 UN: 3089 ADR: 4.1,III Merck Index: 12,05098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...