Lutetium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS
Warning

71-0055-25X25MM 71-0055-50X50MM
100200-488EA 1272.43 USD
100200-488 100201-242
Lutetium ≥99.9%, foil
Lutetium
Formula: Lu
Boiling Pt: 3327 °C
Melting Pt: 1652 °C
Density: 9.842
MDL Number: MFCD00011098
CAS Number: 7439-94-3
EINECS: 231-103-0
Merck Index: 12,05642

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR