Cerium ≥99.9%, foil
Supplier: STREM CHEMICALS INC MS
|
Danger
|
58-0050-25X25MM
58-0050-50X50MM
100198-864EA
484.06
USD
100198-864
100205-970
Cerium ≥99.9%, foil
Cerium
Formula:
Ce MW: 140.12 g/mol Boiling Pt: 3468 Melting Pt: 795 Density: 6.90 |
MDL Number:
MFCD00010924 CAS Number: 7440-45-1 EINECS: 231-154-9 UN: 1333 ADR: 4.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...