Fullerene C70 ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Buckminster-Fullerene C70, [5,6]-Fullerene-C70
06-0603-250MG
06-0603-10MG
06-0603-50MG
100200-378EA
1326.22
USD
100200-378
100207-520
100208-616
Fullerene C70 ≥99%
Fullerene C70
Formula:
C₇₀ MW: 840.77 g/mol Melting Pt: > 280 °C Density: 1.73 g/cm³ (20 °C) |
MDL Number:
MFCD00282904 CAS Number: 115383-22-7 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...