Fullerene C60 ≥99.9% (Buckminsterfullerene)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen
06-0602-25MG
06-0602-100MG
06-0602-500MG
100201-330EA
76.43
USD
100201-330
100204-782
100200-376
Fullerene C60 ≥99.9% (Buckminsterfullerene)
Fullerene C60
Fullerene
Formula:
C₆₀ Melting Pt: > 280 °C Density: ∼1.65 g/cm³ (20 °C) |
MDL Number:
MFCD00151408 CAS Number: 99685-96-8 UN: 1325 ADR: 4.1,III Merck Index: 13,01447 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...