Fullerene C70 ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Buckminster-Fullerene C70, [5,6]-Fullerene-C70

06-0503-10MG 06-0503-250MG 06-0503-50MG
100202-836EA 82.09 USD
100202-836 100207-928 100207-926
Fullerene C70 ≥98%
Fullerene C70

Fullerene

Formula: C₇₀
MW: 840.77 g/mol
Melting Pt: > 280 °C
Density: 1.73 g/cm³ (20 °C)
MDL Number: MFCD00282904
CAS Number: 115383-22-7
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR