Fullerene C60 ≥99.9% (Buckminsterfullerene)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen

06-0502-25MG 06-0502-100MG 06-0502-500MG
100201-326EA 66.52 USD
100201-326 100201-328 100207-518
Fullerene C60 ≥99.9% (Buckminsterfullerene)
Fullerene C60

Fullerene

Formula: C₆₀
Melting Pt: > 280 °C
Density: ∼1.65 g/cm³ (20 °C)
MDL Number: MFCD00151408
CAS Number: 99685-96-8
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=53371479acfcc1403d111c470b60d1af20934c57&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR