Beryllium ≥99.5%, foil

Supplier: STREM CHEMICALS INC MS
Danger

04-0080-50X50MM 04-0080-25X25MM
100200-824EA 2202.34 USD
100200-824 100204-766
Beryllium ≥99.5%, foil
Beryllium
Formula: Be
Boiling Pt: 2970 °C (5 mmHg)
Melting Pt: 1278 °C
Density: 1.85
MDL Number: MFCD00134032
CAS Number: 7440-41-7
EINECS: 231-150-7
UN: 1566
ADR: 6.1,II
Merck Index: 12,01201

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR