Acetone ≥99.5% ACS

Supplier: Spectrum Chemical Mfg. Corp.
Danger

A1020-4LTPL A1020-4LTGL A1020-100ML A1020-1LTGL A1020-200LTB A1020-500MLG
700003-854EA 319.41 USD
700003-854 700003-852 700003-844 700003-846 700003-848 700003-856
Acetone ≥99.5% ACS
Acetone

Acetone, Reagent, ACS is colorless, flammable, liquid. Due to it's micsibilaty with water, it serves as an important solvent. Used as a polar aprotic solvent in a wide range of organic reactions, acetone does not form an azeotrope with water. Thanks to it's low cost and volatility, it is also commonly used to clean laboratory glassware. Another characteristic of Acetone is that it can be cooled with dry ice to -78°C without freezing allowing this solvent to be used to conduct reactions at low temperature. As an ACS grade Reagent, Spectrum Chemical manufactured Acetone is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity

Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.788
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR