Acetanilide ≥99%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
N-Phenylacetamide
A1007-500GM
A1007-50KGBL
A1007-1KG
A1007-250GM
A1007-12KGBL
700007-798EA
275.25
USD
700007-798
700007-800
700007-794
700007-796
700007-792
Acetanilide ≥99%, Reagent Grade
Acetanilide
Acetanilide, Reagent, also known as acetanil, is used as an inhibitor of hydrogen peroxide decomposition as well as to stabilize cellulose ester varnishes. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₈H₉NO MW: 135.17 g/mol Boiling Pt: 304 °C (1013 hPa) Melting Pt: 115 °C Density: 1.22 g/cm³ (20 °C) Flash Pt: 174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008674 CAS Number: 103-84-4 EINECS: 203-150-7 Merck Index: 13,00051 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...