Copper ≥99.999%, wire
Supplier: STREM CHEMICALS INC MS
93-2973-1MT
93-2973-5MT
100204-294EA
141.54
USD
100204-294
100205-500
Copper ≥99.999%, wire
Copper
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.92 Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...