Copper ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS

93-2970-250MM 93-2970-50MM
100207-020EA 111.82 USD
100207-020 100207-022
Copper ≥99.9%, foil
Copper
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.92
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR