Copper(I) chloride, anhydrous ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cuprous chloride, Copper chloride
93-2914-250G
93-2914-1000G
100197-798EA
131.63
USD
100197-798
100211-250
Copper(I) chloride, anhydrous ≥97%
Copper (I) chloride
Formula:
ClCu MW: 99 g/mol Boiling Pt: 1367 Melting Pt: 430 Density: 4.14 Storage Temperature: Ambient |
MDL Number:
MFCD00010971 CAS Number: 7758-89-6 EINECS: 231-842-9 UN: 2802 ADR: 8 III Merck Index: 13,02690 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...