Copper(II) bromide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cupric bromide, Cuprous bromide

93-2908-250G 93-2908-50G
100196-592EA 213.72 USD
100196-592 100210-338
Copper(II) bromide ≥99%
Copper (II) bromide
Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900 °C (1013 hPa)
Melting Pt: 498 °C
Density: 4.77
Storage Temperature: Ambient
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8,III
Merck Index: 13,02656

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR