Nickel ≥99.5%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-2883-500G
93-2883-100G
100205-872EA
322.71
USD
100205-872
100210-332
Nickel ≥99.5%, powder
Nickel
Formula:
Ni Boiling Pt: 2732 °C Melting Pt: 1453 °C Density: 8.90 Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...