Nickel ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-2867-100G 93-2867-500G
100207-828EA 46.71 USD
100207-828 100209-428
Nickel ≥99.9%, powder
Nickel
Formula: Ni
Boiling Pt: 2732 °C
Melting Pt: 1453 °C
Density: 8.90
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR