Iron ≥99%, Powder

Supplier: STREM CHEMICALS INC MS
Danger

93-2663-500G 93-2663-2000G
100204-260EA 100.49 USD
100204-260 100200-090
Iron ≥99%, Powder
Iron
Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000 °C
Melting Pt: 1535 °C
Density: 7,86
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR