Iron ≥99.5%, foil

Supplier: STREM CHEMICALS INC MS
Warning

93-2661-100MM 93-2661-25MM
100199-032EA 331.2 USD
100199-032 100210-302
Iron ≥99.5%, foil
Iron
Formula: Fe
Boiling Pt: 3000 °C
Melting Pt: 1535 °C
Density: 7.86
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR