Vanadium(IV) oxide ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Vanadium dioxide
93-2309-2G
93-2309-50G
100208-526EA
56.62
USD
100208-526
100210-266
Vanadium(IV) oxide ≥99%
Vanadium (IV) oxide
Formula:
O₂V Boiling Pt: 2700 °C (1013 hPa) Melting Pt: 1967 °C Density: 4.339 |
MDL Number:
MFCD00049702 CAS Number: 12036-21-4 EINECS: 234-841-1 UN: 3285 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...