Titanium(IV) bromide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Titanium tetrabromide
93-2221-5G
93-2221-100G
93-2221-25G
100204-232EA
49.54
USD
100204-232
100208-520
100201-078
Titanium(IV) bromide ≥98%
Titanium (IV) bromide
Formula:
Br₄Ti Boiling Pt: 230 °C Melting Pt: 39 °C Density: 2.6 |
MDL Number:
MFCD00011265 CAS Number: 7789-68-6 EINECS: 232-185-0 UN: 1759 ADR: 8,II Merck Index: 12,09617 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...