Titanium(IV) oxide ≥99.99% (Ti basis), PURATREM
Supplier: STREM CHEMICALS INC MS
Synonyms:
Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide
93-2207-25G
93-2207-100G
100199-508EA
268.92
USD
100199-508
100207-464
Titanium(IV) oxide ≥99.99% (Ti basis), PURATREM
Titanium (IV) oxide
Formula:
TiO₂ MW: 79.88 g/mol Boiling Pt: 2900 Melting Pt: 1855 Density: 4.26 Storage Temperature: Ambient |
MDL Number:
MFCD00011269 CAS Number: 13463-67-7 EINECS: 236-675-5 Merck Index: 13,09549 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...