Titanium(IV) butanolate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Titanium (IV) butoxide, Titanium tetrabutanolate, Tetra-n-butyl orthotitanate, Titanium (IV) n-butoxide
93-2204-500G
93-2204-2000G
100200-534EA
96.25
USD
100200-534
100197-744
Titanium(IV) butanolate ≥98%
Titanium(IV) butanolate
Alkoxide, Metal Alkoxides, Volatile Precursors for CVD
Formula:
C₁₆H₃₆O₄Ti Boiling Pt: 312 °C (206 °C (10 mmHg) Melting Pt: –55 °C Density: 0.994 (25°) Flash Pt: 170 °F |
MDL Number:
MFCD00009433 CAS Number: 5593-70-4 EINECS: 227-006-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...