Discandium trioxide ≥99.9% (Sc basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Scandium oxide, Scandium (III) oxide

93-2113-5G 93-2113-25G 93-2113-1G
100210-250EA 210.89 USD
100210-250 100205-408 100197-740
Discandium trioxide ≥99.9% (Sc basis)
Discandium trioxide
Formula: O₃Sc₂
MW: 137.91
Melting Pt: 1000
Density: 3.864
MDL Number: MFCD00011223
CAS Number: 12060-08-1
EINECS: 235-042-0
Merck Index: 14,08392

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR