Calcium carbonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Marble

93-2010-1000G 93-2010-250G
100200-320EA 309.97 USD
100200-320 100205-402
Calcium carbonate ≥98%
Calcium carbonate
Formula: CaCO₃
Melting Pt: 825 °C
Density: 2.930
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR