Potassium fluoride dihydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-1981-50G 93-1981-250G
100205-394EA 43.88 USD
100205-394 100210-230
Potassium fluoride dihydrate ≥98%
Potassium fluoride dihydrate
Formula: FK·2H₂O
Boiling Pt: 1505 °C
Melting Pt: 41 °C
Density: 2.454
MDL Number: MFCD00149929
CAS Number: 13455-21-5
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR