Potassium sulfate ≥99% ACS

Supplier: STREM CHEMICALS INC MS

93-1946-1000G 93-1946-250G
100203-404EA 182.58 USD
100203-404 100204-208
Potassium sulfate ≥99% ACS
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR