Potassium perchlorate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-1935-100G 93-1935-500G
100205-384EA 42.46 USD
100205-384 100199-500
Potassium perchlorate ≥99%
Potassium perchlorate
Formula: ClKO₄
Melting Pt: 610 °C
Density: 2.52
MDL Number: MFCD00011362
CAS Number: 7778-74-7
EINECS: 231-912-9
UN: 1489
ADR: 5.1,II
Merck Index: 13,07738

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR