Potassium chlorate ≥99% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-1913-100G
93-1913-500G
100204-202EA
41.05
USD
100204-202
100209-030
Potassium chlorate ≥99% ACS
Potassium chlorate
Formula:
KClO₃ MW: 122.55 g/mol Boiling Pt: 400 °C (dec.) Melting Pt: 356 °C Density: 2.32 Storage Temperature: Ambient |
MDL Number:
MFCD00011361 CAS Number: 3811-04-9 EINECS: 223-289-7 UN: 1485 ADR: 5.1,II Merck Index: 13,07703 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...