Potassium carbonate ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potash, Carbonic acid potassium salt, Carbonic acid, dipotassium salt

93-1912-500G 93-1912-2000G
100196-522EA 96.25 USD
100196-522 100201-074
Potassium carbonate ≥99% ACS
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Boiling Pt: Dec.
Melting Pt: 891
Density: 2.428
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR