Phenyltrimethoxysilane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Trimethoxy(phenyl)silane

93-1488-250G 93-1488-50G
100210-206EA 254.77 USD
100210-206 100201-924
Phenyltrimethoxysilane ≥97%
Phenyltrimethoxysilane
Formula: C₉H₁₄O₃Si
Boiling Pt: 211 °C
Density: 1,064
Flash Pt: 112 °C
MDL Number: MFCD00025689
CAS Number: 2996-92-1
EINECS: 221-066-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR