Dichloro(methyl)silane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Methyldichlorosilane

93-1486-250G 93-1486-1000G
100198-318EA 111.82 USD
100198-318 100201-068
Dichloro(methyl)silane ≥97%
Dichloro(methyl)silane
Formula: CH₄Cl₂Si
Boiling Pt: 41.5 °C
Melting Pt: –93 °C
Density: 1.110
Flash Pt: –26 °F
MDL Number: MFCD00000494
CAS Number: 75-54-7
EINECS: 200-877-1
UN: 1242
ADR: 4.3,I
REACH: 01-2119436669-21

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR