Silicon(II) oxide ≥99.9% (Si basis) -325 mesh
Supplier: STREM CHEMICALS INC MS
Synonyms:
Silicium (II) oxide, Silicon monoxide, Silicon oxide
93-1478-25G
93-1478-100G
100208-120EA
144.37
USD
100208-120
100208-118
Silicon(II) oxide ≥99.9% (Si basis) -325 mesh
Silicon (II) oxide
Formula:
SiO MW: 44.08 g/mol Boiling Pt: 1880 Melting Pt: 1702 Density: 2.13 Storage Temperature: Ambient |
MDL Number:
MFCD00151536 CAS Number: 10097-28-6 EINECS: 233-232-8 Merck Index: 12,08640 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...