Ammonium hexafluorosilicate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Ammonium silicofluoride Ammonium hexafluorosilicate, Ammonium fluorosilicate, Ammomium fluosilicate

93-1476-2000G 93-1476-500G
100197-708EA 7183.08 USD
100197-708 100201-922
Ammonium hexafluorosilicate ≥98%
Ammonium hexafluorosilicate
Formula: H₈F₆N₂Si
Melting Pt: Dec.
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010887
CAS Number: 16919-19-0
EINECS: 240-968-3
UN: 2854
ADR: 6.1,III
Merck Index: 13,00529

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR